missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Supplier:  TCI America A200425G

 View more versions of this product

Catalog No. A200425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

D-Alanine Benzyl Ester p-Toluenesulfonate
115°C
C17H21NO5S
25 g
NWOPHJSSBMABBD-DDWIOCJRSA-N
4-methylbenzene-1-sulfonic acid; benzyl (2R)-2-aminopropanoate
2802425
≥98.0% (HPLC,T)
41036-32-2
White-Yellow
MFCD00039092
h-d-ala-obzl.tosoh, d-alanine benzyl ester p-toluenesulfonate salt, d-alanine benzyl ester p-toluenesulfonate, d-alanine benzyl ester tosylate, h-d-ala-obzl p-tosylate, h-d-ala-obzl.tos-oh, o-benzyl-d-alanine toluene-p-sulphonate, r-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2r-2-aminopropanoate; para-toluene sulfonate, d-ala-obzltosoh
CC1=CC=C(C=C1)S(O)(=O)=O.C[C@@H](N)C(=O)OCC1=CC=CC=C1
351.42
351.42
Crystalline Powder

Chemical Identifiers

41036-32-2
351.42
NWOPHJSSBMABBD-DDWIOCJRSA-N
2802425
CC1=CC=C(C=C1)S(O)(=O)=O.C[C@@H](N)C(=O)OCC1=CC=CC=C1
C17H21NO5S
MFCD00039092
h-d-ala-obzl.tosoh, d-alanine benzyl ester p-toluenesulfonate salt, d-alanine benzyl ester p-toluenesulfonate, d-alanine benzyl ester tosylate, h-d-ala-obzl p-tosylate, h-d-ala-obzl.tos-oh, o-benzyl-d-alanine toluene-p-sulphonate, r-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2r-2-aminopropanoate; para-toluene sulfonate, d-ala-obzltosoh
4-methylbenzene-1-sulfonic acid; benzyl (2R)-2-aminopropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions