missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Alanine benzyl ester p-toluenesulfonate, 98%

Manufacturer:  Alfa Aesar™ H6241214

 View more versions of this product

Catalog No. AAH6241214



Specifications

Specifications

D-Alanine benzyl ester p-toluenesulfonate
C17H21NO5S
h-d-ala-obzl.tosoh, d-alanine benzyl ester p-toluenesulfonate salt, d-alanine benzyl ester p-toluenesulfonate, d-alanine benzyl ester tosylate, h-d-ala-obzl p-tosylate, h-d-ala-obzl.tos-oh, o-benzyl-d-alanine toluene-p-sulphonate, r-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2r-2-aminopropanoate; para-toluene sulfonate, d-ala-obzltosoh
CC1=CC=C(C=C1)S(=O)(=O)O.CC(C(=O)OCC1=CC=CC=C1)N
351.417
351.42
25g
41036-32-2
MFCD00039092
NWOPHJSSBMABBD-DDWIOCJRSA-N
benzyl (2R)-2-aminopropanoate;4-methylbenzenesulfonic acid
2802425
98%

Chemical Identifiers

41036-32-2
351.417
NWOPHJSSBMABBD-DDWIOCJRSA-N
2802425
CC1=CC=C(C=C1)S(=O)(=O)O.CC(C(=O)OCC1=CC=CC=C1)N
C17H21NO5S
MFCD00039092
h-d-ala-obzl.tosoh, d-alanine benzyl ester p-toluenesulfonate salt, d-alanine benzyl ester p-toluenesulfonate, d-alanine benzyl ester tosylate, h-d-ala-obzl p-tosylate, h-d-ala-obzl.tos-oh, o-benzyl-d-alanine toluene-p-sulphonate, r-benzyl 2-aminopropanoate 4-methylbenzenesulfonate, benzyl 2r-2-aminopropanoate; para-toluene sulfonate, d-ala-obzltosoh
benzyl (2R)-2-aminopropanoate;4-methylbenzenesulfonic acid
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C

Documents