missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Alanine, 99%

Manufacturer:  Alfa Aesar™ A1023106

 View more versions of this product

Catalog No. AAA1023106



Description

Description

Applications
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.

Solubility
Soluble in water (155 mg/ml at 20°C), ethanol (slightly), and pyridine (slightly). Insoluble in ether, and acetone.

Notes
Store in cool, dry conditions in well sealed containers. Strong oxidizing agents.
Specifications

Specifications

D-Alanine
C3H7NO2
1720249
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
CC(C(=O)O)N
89.094
CHEBI:15570
99%
5g
338-69-2
MFCD00008077
14,204
QNAYBMKLOCPYGJ-UWTATZPHSA-N
(2R)-2-aminopropanoic acid
71080
89.09
~295°C (decomposition)
−14° (c=5 in 5N HCl)

Chemical Identifiers

338-69-2
89.094
QNAYBMKLOCPYGJ-UWTATZPHSA-N
71080
(2R)-2-aminopropanoic acid
C3H7NO2
MFCD00008077
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
CHEBI:15570
CC(C(=O)O)N
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents