missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Alaninamide hydrochloride, 98%

$45.18 - $178.89

Chemical Identifiers

CAS 71810-97-4
Molecular Formula C3H9ClN2O
Molecular Weight (g/mol) 124.568
MDL Number MFCD00039093
InChI Key FIAINKIUSZGVGX-HSHFZTNMSA-N
Synonym r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl
PubChem CID 2775814
IUPAC Name (2R)-2-aminopropanamide;hydrochloride
SMILES CC(C(=O)N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6353503 View Documents Alfa Aesar™
H6353503
1g Each for $45.18
 
AAH6353506 View Documents Alfa Aesar™
H6353506
5g Each for $178.89
 
Description

Description

Applications
D-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in 1% acetic acid and water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

71810-97-4
124.568
FIAINKIUSZGVGX-HSHFZTNMSA-N
2775814
CC(C(=O)N)N.Cl
C3H9ClN2O
MFCD00039093
r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl
(2R)-2-aminopropanamide;hydrochloride

Specifications

71810-97-4
MFCD00039093
FIAINKIUSZGVGX-HSHFZTNMSA-N
(2R)-2-aminopropanamide;hydrochloride
2775814
Powder
White
D-Alaninamide hydrochloride
C3H9ClN2O
r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl
CC(C(=O)N)N.Cl
124.568
124.57
98%
1g
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning