Learn More
D-Alaninamide hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6353506
Description
D-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
Solubility
Soluble in 1% acetic acid and water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
D-Alaninamide hydrochloride | |
White | |
MFCD00039093 | |
Soluble in 1% acetic acid and water. | |
CC(C(=O)N)N.Cl | |
124.568 | |
124.57 | |
5 g |
71810-97-4 | |
C3H9ClN2O | |
r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl | |
FIAINKIUSZGVGX-HSHFZTNMSA-N | |
(2R)-2-aminopropanamide;hydrochloride | |
2775814 | |
98% | |
Powder |
Chemical Identifiers
71810-97-4 | |
124.568 | |
FIAINKIUSZGVGX-HSHFZTNMSA-N | |
2775814 | |
CC(C(=O)N)N.Cl |
C3H9ClN2O | |
MFCD00039093 | |
r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl | |
(2R)-2-aminopropanamide;hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only