Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Ala&superscript_1;] Peptide T, amide

For cellular and molecular biology applications

Specifications

Product Type [D-Ala1] Peptide T, amide
Molecular Weight 856.89
Storage Requirements -30° to -10°C
Sequence D-Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66424LB0

 
alfa aesar™
J66424LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66424MCR

 
alfa aesar™
J66424MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [D-Ala1] Peptide T, amide
Molecular Weight 856.89
Storage Requirements -30° to -10°C
Sequence D-Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC