Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Ala&superscript_2;, Hyp&superscript_4;, Tyr&superscript_5;] beta-Casomorphin (1-5)Amide

For cellular and molecular biology applications

Specifications

Product Type [D-Ala2, Hyp4, Tyr5] beta-Casomorphin (1-5) amide
Molecular Weight 673.73
Storage Requirements -30° to -10°C
Sequence Tyr-D-Ala-Phe-Hyp-Tyr-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66290LB0

 
alfa aesar™
J66290LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66290MB

 
alfa aesar™
J66290MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [D-Ala2, Hyp4, Tyr5] beta-Casomorphin (1-5) amide
Molecular Weight 673.73
Storage Requirements -30° to -10°C
Sequence Tyr-D-Ala-Phe-Hyp-Tyr-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC