Please login to your online account to display your discounted pricing

D-64131, Tocris

Inhibitor of tubulin polymerization; Antitumor in vivo

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material D-64131
Assay Percent Range >99%
Molecular Formula C16H13NO2
Formula Weight 251.28
Synonym (5-Methoxy-1H-indol-2-yl)phenylmethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

164310

 
r & d systems
164310
10mg Each for $201.36

164350

 
r & d systems
164350
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 251.28
Quantity 10mg
Synonym (5-Methoxy-1H-indol-2-yl)phenylmethanone
Assay Percent Range >99%
Molecular Formula C16H13NO2
Chemical Name or Material D-64131
CAS 74588-78-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature