Please login to your online account to display your discounted pricing

Alfa Aesar™ D-3-Bromocamphor-8-sulfonic acid ammonium salt, 98%

$61.64 - $736.81

Chemical Identifiers

Chemical Name or Material D-3-Bromocamphor-8-sulfonic acid ammonium salt
CAS 14575-84-9
Molecular Formula C10H18BrNO4S
Formula Weight 328.23
MDL Number MFCD00167438
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1988606

  SDS
alfa aesar™
A1988606
5g Each for $61.64

AAA1988614

  SDS
alfa aesar™
A1988614
25g Each for $219.99

AAA1988622

  SDS
alfa aesar™
A1988622
100g Each for $736.81
Description & Specifications

Specifications

Formula Weight 328.23
Chemical Name or Material D-3-Bromocamphor-8-sulfonic acid ammonium salt
Assay 98%
Molecular Formula C10H18BrNO4S
MDL Number MFCD00167438
CAS 14575-84-9
Melting Point ≈284°C (dec.)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
D-3-Bromocamphor-8-sulfonic acid ammonium salt is used in optical resolutions. It is also used as pharmaceutical intermediates.

Solubility
Soluble in Methanol and Water (200 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at -20°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.