Please login to your online account to display your discounted pricing

Alfa Aesar™ D-3-Bromocamphor-10-sulfonic acid monohydrate, 99%

Chemical Identifiers

Chemical Name or Material D-3-Bromocamphor-10-sulfonic acid monohydrate
CAS 209736-59-4
Molecular Formula C10H15BrO4S·H2O
Formula Weight 329.22g/mol (311.20g/mol Anhydrous)
MDL Number MFCD00209633
UN Number UN1759
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1679106

 
alfa aesar™
A16791-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 115° to 119°C
Quantity 5g
Chemical Name or Material D-3-Bromocamphor-10-sulfonic acid monohydrate
CAS 209736-59-4
Assay 99%
Molecular Formula C10H15BrO4S·H2O
Formula Weight 329.22g/mol (311.20g/mol Anhydrous)
MDL Number MFCD00209633
UN Number UN1759
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.