Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(-)-2-Phenylglycine chloride hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material D-(-)-2-Phenylglycine chloride hydrochloride
CAS 39878-87-0
Formula Weight 206.07
MDL Number MFCD00039094
UN Number UN3261
Beilstein 5405183
Synonym (R)-α-Aminophenylacetyl chloride hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5682803

 
alfa aesar™
H5682803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5682806

 
alfa aesar™
H5682806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.07
Chemical Name or Material D-(-)-2-Phenylglycine chloride hydrochloride
TSCA TSCA
Physical Form Powder
CAS 39878-87-0
Beilstein 5405183
Synonym (R)-α-Aminophenylacetyl chloride hydrochloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Color Pale brown
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number CY1920000
Assay 98%
MDL Number MFCD00039094
UN Number UN3261
Melting Point 177°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.