Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(-)-2-Cyclohexylglycine, 95%

Chemical Identifiers

Chemical Name or Material D-(-)-2-Cyclohexylglycine
CAS 14328-52-0
Formula Weight 157.21
MDL Number MFCD01311678
Beilstein 3196806
Synonym H-D-Chg-OH; (R)-(+)-α-Aminocyclohexaneacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6338103

  SDS
alfa aesar™
H6338103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6338106

  SDS
alfa aesar™
H6338106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6338114

  SDS
alfa aesar™
H6338114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.21
Physical Form Solid
MDL Number MFCD01311678
Beilstein 3196806
Assay 95%
CAS 14328-52-0
Chemical Name or Material D-(-)-2-Cyclohexylglycine
Synonym H-D-Chg-OH; (R)-(+)-α-Aminocyclohexaneacetic acid
Quantity 1g