missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(-)-2-Cyclohexylglycine, 95%

Manufacturer:  Alfa Aesar™ H6338114

 View more versions of this product

Catalog No. AAH6338114



Specifications

Specifications

D-(-)-2-Cyclohexylglycine
C8H15NO2
3196806
WAMWSIDTKSNDCU-SSDOTTSWSA-N
(2R)-2-amino-2-cyclohexylacetic acid
736849
Solid
25g
14328-52-0
MFCD01311678
d-cyclohexylglycine, d-alpha-cyclohexylglycine, r-2-amino-2-cyclohexylacetic acid, h-d-chg-oh, 2-cyclohexyl-d-glycine, 2r-amino cyclohexyl acetic acid, 2r-2-amino-2-cyclohexylacetic acid, d-2-cyclohexylglycine, d-cyclohexyl glycine, h-cyclohexyl-d-gly-oh
C1CCC(CC1)C(C(=O)O)N
157.213
157.21
95%

Chemical Identifiers

14328-52-0
157.213
WAMWSIDTKSNDCU-SSDOTTSWSA-N
736849
C1CCC(CC1)C(C(=O)O)N
C8H15NO2
MFCD01311678
d-cyclohexylglycine, d-alpha-cyclohexylglycine, r-2-amino-2-cyclohexylacetic acid, h-d-chg-oh, 2-cyclohexyl-d-glycine, 2r-amino cyclohexyl acetic acid, 2r-2-amino-2-cyclohexylacetic acid, d-2-cyclohexylglycine, d-cyclohexyl glycine, h-cyclohexyl-d-gly-oh
(2R)-2-amino-2-cyclohexylacetic acid
SDS Documents