missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(-)-2-Aminobutyric Acid, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152140250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D(-)-2-Aminobutyric acid | |
Authentic | |
Glass bottle | |
C2H5CH(NH2)CO2H | |
25g | |
−21.20° (20°C c=2,6N HCl) | |
d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid | |
QWCKQJZIFLGMSD-GSVOUGTGSA-N | |
(2R)-2-aminobutanoic acid | |
439691 | |
103.12 |
2623-91-8 | |
99+% | |
C4H9NO2 | |
MFCD00064414 | |
04, 408 | |
-21.2 | |
Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol,insoluble in ether | |
CC[C@@H](N)C(O)=O | |
103.12 | |
CHEBI:28797 | |
99+% |
Chemical Identifiers
2623-91-8 | |
103.12 | |
QWCKQJZIFLGMSD-GSVOUGTGSA-N | |
439691 | |
(2R)-2-aminobutanoic acid |
C4H9NO2 | |
MFCD00064414 | |
d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid | |
CHEBI:28797 | |
CC[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 220-084-4
RUO â Research Use Only