Please login to your online account to display your discounted pricing

D(-)-2-Aminobutyric Acid, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material D(-)-2-Aminobutyric acid
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C4H9NO2
Linear Formula C2H5CH(NH2)CO2H
Formula Weight 103.12
MDL Number MFCD00064414
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC152140250

  SDS
thermo fisher scientific
AC152140250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC152140050

  SDS
fisher scientific acros
152140050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥99.0%
Formula Weight 103.12
Quantity 25g
Color White
Specific Rotation Condition −21.20° (20°C c=2,6N HCl)
Beilstein 04, 408
MDL Number MFCD00064414
CAS 2623-91-8
Chemical Name or Material D(-)-2-Aminobutyric acid
Packaging Glass bottle
Molecular Formula C4H9NO2
Linear Formula C2H5CH(NH2)CO2H
Infrared Spectrum Authentic
Specific Rotation -20.2° to -22.2° (20°C, 589nm) c=2, 6N HCl
Assay Percent Range 99+%