missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(-)-2-Aminobutyric Acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 152140050

 View more versions of this product

Catalog No. AC152140050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

D(-)-2-Aminobutyric acid
Glass bottle
White
5g
99+%
C2H5CH(NH2)CO2H
04, 408
Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol,insoluble in ether
CCC(C(=O)O)N
103.12
CHEBI:28797
Crystalline Powder
Authentic
-21.2
>300°C
2623-91-8
C4H9NO2
MFCD00064414
2r-2-aminobutanoic acid
QWCKQJZIFLGMSD-GSVOUGTGSA-N
(2R)-2-aminobutanoic acid
439691
103.12
99+%

Chemical Identifiers

2623-91-8
103.12
QWCKQJZIFLGMSD-GSVOUGTGSA-N
439691
(2R)-2-aminobutanoic acid
C4H9NO2
MFCD00064414
2r-2-aminobutanoic acid
CHEBI:28797
CCC(C(=O)O)N
Safety and Handling

Safety and Handling

EINECSNumber : 220-084-4

SDS Documents