missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-(-)-2-Aminobutyric acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1409606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D-(-)-2-Aminobutyric acid | |
>300°C | |
MFCD00064414 | |
1720934 | |
QWCKQJZIFLGMSD-GSVOUGTGSA-N | |
CC[C@@H](N)C(O)=O | |
103.12 | |
CHEBI:28797 | |
≥98% |
2623-91-8 | |
C4H9NO2 | |
5 g | |
d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid | |
−20.5° (c=4.8 in 6N HCl) | |
(2R)-2-aminobutanoic acid | |
439691 | |
103.12 |
Chemical Identifiers
2623-91-8 | |
103.12 | |
QWCKQJZIFLGMSD-GSVOUGTGSA-N | |
439691 | |
(2R)-2-aminobutanoic acid |
C4H9NO2 | |
MFCD00064414 | |
d-2-aminobutyric acid, r-2-aminobutanoic acid, d---2-aminobutyric acid, 2r-2-aminobutanoic acid, d-alpha-aminobutyric acid, r---2-aminobutyric acid, h-d-abu-oh, d--2-aminobutyric acid, d-2-aminobutyrate, d-2-aminobutanoic acid | |
CHEBI:28797 | |
CC[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 220-084-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only