Please login to your online account to display your discounted pricing

Alfa Aesar™ D-2-Aminoadipic acid, 98%

$147.22 - $1,482.65

Chemical Identifiers

Chemical Name or Material D-2-Aminoadipic acid
CAS 7620-28-2
Molecular Formula C6H11NO4
Formula Weight 161.16
MDL Number MFCD00063118
Beilstein 1724347
Synonym H-D-2-Aad-OH; (R)-2-Aminohexanedioic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2734503

  SDS
alfa aesar™
H2734503
1g Each for $459.57

AAH2734506

  SDS
alfa aesar™
H2734506
5g Each for $1,482.65

AAH27345MD

  SDS
alfa aesar™
H27345MD
250mg Each for $147.22
Description & Specifications

Specifications

Formula Weight 161.16
CAS 7620-28-2
Synonym H-D-2-Aad-OH; (R)-2-Aminohexanedioic acid
Assay 98%
Quantity 1g
Melting Point 205°C
Chemical Name or Material D-2-Aminoadipic acid
Beilstein 1724347
Molecular Formula C6H11NO4
MDL Number MFCD00063118