missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-2-Amino-3-phenyl-1-propanol, 98%, Thermo Scientific™

$406.30 - $406.30

Chemical Identifiers

CAS 5267-64-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00064298
Synonym +-d-phenylalaninol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, d-+-phenylalaninol, d-penylalaninol, d-phenylalaninol, r-+-2-amino-3-phenyl-1-propanol, r-2-amino-3-phenyl-1-propanol, r-2-amino-3-phenylpropan-1-ol, r-phenylalaninol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC417340010
SDS View Documents Promotion Details
Thermo Fisher Scientific
417340010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC417340100
SDS View Documents Promotion Details
Thermo Fisher Scientific
417340100
10g
Each for $406.30
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5267-64-1
151.21
+-d-phenylalaninol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, d-+-phenylalaninol, d-penylalaninol, d-phenylalaninol, r-+-2-amino-3-phenyl-1-propanol, r-2-amino-3-phenyl-1-propanol, r-2-amino-3-phenylpropan-1-ol, r-phenylalaninol
C9H13NO
MFCD00064298

Specifications

5267-64-1
90°C to 94°C
Authentic
MFCD00064298
+-d-phenylalaninol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, d-+-phenylalaninol, d-penylalaninol, d-phenylalaninol, r-+-2-amino-3-phenyl-1-propanol, r-2-amino-3-phenyl-1-propanol, r-2-amino-3-phenylpropan-1-ol, r-phenylalaninol
Solubility in water: soluble. Other solubilities: soluble in ethanol,benzene,chloroform
151.21
Crystalline Powder and Chunks
D(+)-2-Amino-3-phenyl-1-propanol, 98%
White to Yellow
97.5% min. (HPLC)
C9H13NO
Glass bottle
1g
+ 23
151.21
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 226-086-1

SDS
Product Certifications

RUO – Research Use Only