Please login to your online account to display your discounted pricing

D(+)-10-Camphorsulfonyl chloride, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  204631000

 View more versions of this product

Catalog No. AC204631000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 250.74
Quantity 100g
Color Off-white to beige
EINECS Number 244-314-8
Melting Point 63° to 68°C
Beilstein 11, 316
Fieser 01,109; 15,68
CAS 21286-54-4
Health Hazard 1 Danger
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Specific Rotation +30° to +36° (22°C, 589nm) (c=1, CHCl3)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material D(+)-10-Camphorsulfonyl chloride
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H15ClO3S
MDL Number MFCD00064156
Infrared Spectrum Authentic

Specifications

Chemical Name or Material D(+)-10-Camphorsulfonyl chloride
Name Note 97%
CAS 21286-54-4
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H15ClO3S
Formula Weight 250.74
MDL Number MFCD00064156
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents