Learn More
D(+)-10-Camphorsulfonyl chloride, 97%, ACROS Organics™
$186.15 - $186.15
Chemical Identifiers
CAS | 21286-54-4 |
---|---|
Molecular Formula | C10H15ClO3S |
Molecular Weight (g/mol) | 250.74 |
MDL Number | MFCD00064156 |
InChI Key | BGABKEVTHIJBIW-GMSGAONNSA-N |
Synonym | +-camphor-10-sulfonic acid chloride, +-camphor-10-sulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-10-camphorsulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, camphor-10-sulfonyl chloride, d +-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride |
PubChem CID | 88856 |
IUPAC Name | [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride |
SMILES | CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC204630050
|
Thermo Fisher Scientific
204630050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC204630250
|
Thermo Fisher Scientific
204630250 |
25g | Glass bottle |
Each for $186.15
|
|
AC204631000
|
Thermo Fisher Scientific
204631000 |
100g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
21286-54-4 | |
250.74 | |
BGABKEVTHIJBIW-GMSGAONNSA-N | |
88856 | |
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C |
C10H15ClO3S | |
MFCD00064156 | |
+-camphor-10-sulfonic acid chloride, +-camphor-10-sulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-10-camphorsulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, camphor-10-sulfonyl chloride, d +-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride | |
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride |
Specifications
Authentic | |
+ 33.00 | |
63.0°C to 68.0°C | |
21286-54-4 | |
C10H15ClO3S | |
11, 316 | |
+-camphor-10-sulfonic acid chloride, +-camphor-10-sulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-10-camphorsulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, camphor-10-sulfonyl chloride, d +-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride | |
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C | |
250.74 | |
250.74 | |
97% |
Glass bottle | |
White to Beige | |
5g | |
96% min. (ex Cl) (Argentometry) | |
MFCD00064156 | |
01,109; 15,68 | |
BGABKEVTHIJBIW-GMSGAONNSA-N | |
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride | |
88856 | |
Crystalline Powder | |
D(+)-10-Camphorsulfonyl chloride, 97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 244-314-8