Please login to your online account to display your discounted pricing

CZC 54252 hydrochloride, Tocris™

Potent LRRK2 inhibitor; neuroprotective

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material CZC 54252 hydrochloride
Assay Percent Range >98%
Molecular Formula C22H25ClN6O4S·HCl
Formula Weight 541.45
Synonym N-[-[[5-Chloro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]methanesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

453450

 
r & d systems
RDS453450
50mg Each for $1,116.26

453410

 
r & d systems
RDS453410
10mg Each for $265.15
Description & Specifications

Specifications

Chemical Name or Material CZC 54252 hydrochloride
Quantity 50mg
CAS 1191911-27-9
Recommended Storage Store at +4°C
Assay Percent Range >98%
Molecular Formula C22H25ClN6O4S·HCl
Formula Weight 541.45
Synonym N-[-[[5-Chloro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]methanesulfonamide hydrochloride
Safety and Handling

Store at +4°C