Please login to your online account to display your discounted pricing

CZC 24832, Tocris™

Selective inhibitor of PI 3-kinase γ

$330.06 - $1,362.21

Chemical Identifiers

Chemical Name or Material CZC 24832
Assay Percent Range >98%
Molecular Formula C15H17FN6O2S
Formula Weight 364.4
Synonym 5-(2-Amino-8-fluoro[1,2,4]triazolo[1,5-a]pyridin-6-yl)-N-(1,1-dimethylethyl)-3-pyridinesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467410

 
r & d systems
RDS467410
10mg Each for $330.06

467450

 
r & d systems
RDS467450
50mg Each for $1,362.21
Description & Specifications

Specifications

Formula Weight 364.4
Quantity 10mg
Synonym 5-(2-Amino-8-fluoro[1,2,4]triazolo[1,5-a]pyridin-6-yl)-N-(1,1-dimethylethyl)-3-pyridinesulfonamide
Chemical Name or Material CZC 24832
Assay Percent Range >98%
Molecular Formula C15H17FN6O2S
CAS 1159824-67-5
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C