Please login to your online account to display your discounted pricing

Cytosine-β-D-arabinofuranoside Hydrochloride, Fisher BioReagents


Quantity 100mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2512100

 View more versions of this product

Catalog No. BP2512100

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 197°C
Physical Form Powder/Solid
Formula Weight 279.7
Quantity 100mg
Molecular Formula C9H12N3O5 · HCl
Color White
Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
Lambda Max 174 to 286nm
Identification Pass Test
Packaging Amber Glass
CAS 69-74-9
FTIR Conforms to reference
Recommended Storage 4°C
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
CAS 69-74-9
Physical Form Powder/Solid
Molecular Formula C9H12N3O5 · HCl
Formula Weight 279.7
Accessories
 

4°C

SDS & Safety Documents