Please login to your online account to display your discounted pricing

Cytosine-β-D-arabinofuranoside Hydrochloride, Fisher BioReagents


Quantity 1g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP25121

 View more versions of this product

Catalog No. BP25121

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

FTIR Conforms to reference
Identification Pass Test
Lambda Max 174 to 286nm
Packaging Amber Glass
Color White
Melting Point 197°C / °F
Quantity 1g
Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
CAS 69-74-9
Physical Form Powder; Solid
Molecular Formula C9H12N3O5 · HCl
Formula Weight 279.7g/mol
ChemAlert Storage Symbol Gray
Recommended Storage 4°C

Specifications

Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
CAS 69-74-9
Physical Form Powder; Solid
Molecular Formula C9H12N3O5 · HCl
Formula Weight 279.7g/mol

Cytosine-β-D-Arabinofuranoside Hydrochloride inhibits incorporation of labeled thymidine into DNA.

Accessories
 

4°C

SDS & Safety Documents