Please login to your online account to display your discounted pricing

Cytosine-β-D-arabinofuranoside Hydrochloride, Fisher BioReagents

$130.02 - $130.02

Chemical Identifiers

Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
CAS 69-74-9
Physical Form Powder/Solid
Molecular Formula C9H12N3O5 · HCl
Formula Weight 279.7
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP2512100

  SDS
fisher scientific company
BP2512100
Amber Glass 100mg Each for $130.02
Case of 5 Each for $650.10
Description & Specifications

Specifications

Melting Point 197°C
Physical Form Powder/Solid
Formula Weight 279.7
Quantity 100mg
Molecular Formula C9H12N3O5 · HCl
Color White
Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
Lambda Max 174 to 286nm
Identification Pass Test
Packaging Amber Glass
CAS 69-74-9
FTIR Conforms to reference
Recommended Storage 4°C
ChemAlert Storage Symbol Gray

Cytosine-β-D-Arabinofuranoside Hydrochloride inhibits incorporation of labeled thymidine into DNA.

Safety and Handling
 

4°C