missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytosine β-D-arabinofuranoside hydrochloride, 99%, crystalline, ACROS Organics™
$1,331.70 - $1,331.70
Chemical Identifiers
CAS | 69-74-9 |
---|---|
Molecular Formula | C9H13N3O5·HCl |
Molecular Weight (g/mol) | 279.67 |
MDL Number | MFCD00012839 |
InChI Key | KCURWTAZOZXKSJ-JCEIVDPDSA-N |
Synonym | cytarabine hydrochloride |
PubChem CID | 134129637 |
IUPAC Name | 4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride |
SMILES | C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC227180050
|
Thermo Fisher Scientific
AC227180050 |
5g | Glass bottle |
Each for $1,331.70
|
|
Chemical Identifiers
69-74-9 | |
279.67 | |
KCURWTAZOZXKSJ-JCEIVDPDSA-N | |
134129637 | |
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl |
C9H13N3O5·HCl | |
MFCD00012839 | |
cytarabine hydrochloride | |
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride |
Specifications
Authentic | |
+127° to +133° (20°C, 589nm) ((c=0.7, H2O)) | |
197.0°C to 198.0°C | |
69-74-9 | |
C9H13N3O5·HCl | |
10, 2778 | |
KCURWTAZOZXKSJ-JCEIVDPDSA-N | |
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride | |
134129637 | |
Crystalline Powder | |
Cytosine β-D-arabinofuranoside hydrochloride |
Glass bottle | |
White | |
5g | |
98.5% min. (HPLC) | |
MFCD00012839 | |
cytarabine hydrochloride | |
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl | |
279.67 | |
279.67 | |
99% |
Safety and Handling
Causes serious eye irritation, Suspected of causing genetic defects, Suspected of damaging the unborn child, May cause an allergic skin reaction
Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Skin sensitisation (category 1)
EINECSNumber : 200-713-9
RMF - Research Use or Further Mfg