missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytosine β-D-arabinofuranoside hydrochloride, 99%, crystalline, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC227180050

 View more versions of this product

Catalog No. AC227180050



Specifications

Specifications

Cytosine β-D-arabinofuranoside hydrochloride
Glass bottle
White
5g
98.5% min. (HPLC)
MFCD00012839
cytarabine hydrochloride
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl
279.67
279.67
99%
Authentic
+127° to +133° (20°C, 589nm) ((c=0.7, H2O))
197.0°C to 198.0°C
69-74-9
C9H13N3O5·HCl
10, 2778
KCURWTAZOZXKSJ-JCEIVDPDSA-N
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
134129637
Crystalline Powder

Chemical Identifiers

69-74-9
279.67
KCURWTAZOZXKSJ-JCEIVDPDSA-N
134129637
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl
C9H13N3O5·HCl
MFCD00012839
cytarabine hydrochloride
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of damaging the unborn child.

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-713-9

SDS Documents