missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin, Multi Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin, Multi Ab-1, Mouse Monoclonal Antibody.

$238.63 - $694.82

Specifications

Antigen Cytokeratin, Multi Ab-1
Clone C-11
Applications Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS149P Thermo Scientific™ Lab Vision™
MS149P
Purified Ab with BSA and Azide 1mL Each for $592.41
MS149P0 Thermo Scientific™ Lab Vision™
MS149P0
Purified Ab with BSA and Azide 100μL Each for $238.63
MS149P1 Thermo Scientific™ Lab Vision™
MS149P1
Purified Ab with BSA and Azide 500μL Each for $406.69
MS149P1ABX Thermo Scientific™ Lab Vision™
MS149P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $528.88
MS149PABX Thermo Scientific™ Lab Vision™
MS149PABX
Ab without BSA/Azide (Frozen) 200μL Each for $694.82
MS149R7 Thermo Scientific
MS149R7
Ready-to-use for IHC (UVLP) 7mL Each for $342.76
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin, Multi Ab-1, Mouse Monoclonal Antibody.

Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: C-11

Isotype: IgG1

Species Reactivity: Human, Cow, Rat, Mouse and Frog. Others not known.

Epitope: Not determined

Immunogen: Cytokeratin-enriched preparation from cultured human A431

Positive Control: Skin or colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytokeratin, Multi Ab-1
Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Unlabeled
RUO
Primary
IgG1
C-11
Monoclonal
Murine
Cytoskeletal Protein
Keratin-enriched preparation from cultured human A431