missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin, Multi Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin, Multi Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS149P1ABX

 View more versions of this product

Catalog No. MS149P1ABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin, Multi Ab-1, Mouse Monoclonal Antibody.

Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: C-11

Isotype: IgG1

Species Reactivity: Human, Cow, Rat, Mouse and Frog. Others not known.

Epitope: Not determined

Immunogen: Cytokeratin-enriched preparation from cultured human A431

Positive Control: Skin or colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytokeratin, Multi Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1
RUO
Primary
Bovine,Frog,Human,Murine,Rat
Immunofluorescence,Immunohistochemistry (Paraffin)
C-11
Unlabeled
Concentrated
Keratin-enriched preparation from cultured human A431
100μL
Cytoskeletal Protein
Monoclonal