Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin, High Molecular Weight Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

$229.67 - $786.61

Specifications

Antigen Cytokeratin, High Molecular Weight Ab-3
Clone 34BetaE12
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1447R7

 
thermo scientific™ lab vision™
MS1447R7
7mL Each for $459.37

MS1447RQ

 
thermo scientific™ lab vision™
MS1447RQ
12mL Each for $693.98

MS1447S

 
thermo scientific™ lab vision™
MS1447S
1mL Each for $786.61

MS1447S0

 
thermo scientific
MS1447S0
100µL Each for $229.67

MS1447S1

 
thermo scientific™ lab vision™
MS1447S1
500µL Each for $649.39
Description & Specifications

Specifications

Antigen Cytokeratin, High Molecular Weight Ab-3
Clone 34BetaE12
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Mouse
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen Solubilized keratin extracted from human stratum corneum

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Cytokeratin, High Molecular Weight Ab-3, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

In normal cells, Ab-3 labels squamous, ductal and other complex epithelia. It reacts with benign small-acinir lesions of the prostate and does not react with hepatocytes, pancreatic acinar cells, proximal renal tubes or endometrial glands. In tumor cells, this antibody is reactive with both squamous and ductal neoplasms and variably with those derived from simple epithelium.

Host Species: Mouse

Clone: 34βE12

Isotype: IgG1

Species Reactivity: Human, Monkey, Horse and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Solubilized keratin extracted from human stratum corneum

Molecular Weight: 68 kDa (Cytokeratin 1), 58 kDa (Cytokeratin 5), 56.5 kDa (Cytokeratin 10) and 50 kDa (Cytokeratin 14)

Positive Control: Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence