Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 8/18 Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 8/18 Ab-2, Mouse Monoclonal Antibody.

$808.27 - $808.27

Specifications

Antigen Cytokeratin 8/18 Ab-2
Clone K8.8 + DC10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1603PABX

 
thermo scientific™ lab vision™
MS1603PABX
Ab without BSA/Azide (Frozen) 200µL Each for $808.27
Description & Specifications

Specifications

Antigen Cytokeratin 8/18 Ab-2
Clone K8.8 + DC10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen Semi-purified human cytokeratin preparation

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 8/18 Ab-2, Mouse Monoclonal Antibody.

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).

Host Species: Mouse

Clone: K8.8 + DC10

Isotype: IgG1/κ + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Semi-purified human cytokeratin preparation

Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)

Positive Control: Skin or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)