missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytokeratin 7 Mouse anti-Human, eFluor 615, Clone: LP5K, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $339.30

Specifications

Antigen Cytokeratin 7
Clone LP5K
Host Species Murine
Species Reactivity Human
Applications Immunocytochemistry,Immunohistochemistry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245829 Life Technologies
LS42900580
25μg Each for $160.95
50245830 Life Technologies
LS42900582
100μg Each for $339.30
Specifications

Description

This LP5K monoclonal antibody reacts with human cytokeratin 7 (K7), a 54kDa type II (or basic) keratin expressed either alone or paired with cytokeratin 19 in simple epithelia, mesothelium, urothelium, and pseudostratified epithelium. Expression of cytokeratin 7 in the gastric foveolar, intestinal, and stratified squamous epithelia is extremely low or undetectable. Cytokeratins form the intracellular cytoskeletal network that maintains the integrity and stability of cells and tissues. In addition, most carcinomas express cytokeratin 7. The coordinated expression of this keratin with cytokeratin 20 is commonly used as a diagnostic marker for a variety of carcinomas.
Description & Specifications

Specifications

Cytokeratin 7
Murine
Immunocytochemistry,Immunohistochemistry
eFluor 615
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
LP5K
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer