Please login to your online account to display your discounted pricing

Cytokeratin 7 Mouse anti-Human, Clone: LP5K, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $259.55

Specifications

Antigen Cytokeratin 7
Clone LP5K
Host Species Mouse
Gene Alias keratin 7
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013765

 
affymetrix
14900582
100μg Each for $259.55

5013764

 
affymetrix
14900580
25μg Each for $129.05
Description & Specifications

Specifications

Antigen Cytokeratin 7
Clone LP5K
Host Species Mouse
Gene Alias keratin 7
Species Reactivity Human
Applications Immunocytochemistry
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This LP5K monoclonal antibody reacts with human cytokeratin 7 (K7), a 54kDa type II (or basic) keratin expressed either alone or paired with cytokeratin 19 in simple epithelia, mesothelium, urothelium, and pseudostratified epithelium. Expression of cytokeratin 7 in the gastric foveolar, intestinal, and stratified squamous epithelia is extremely low or undetectable. Cytokeratins form the intracellular cytoskeletal network that maintains the integrity and stability of cells and tissues. In addition, most carcinomas express cytokeratin 7. The coordinated expression of this keratin with cytokeratin 20 is commonly used as a diagnostic marker for a variety of carcinomas.