Please login to your online account to display your discounted pricing

Cytokeratin 6 Mouse anti-Human, Clone: LL020, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $323.35

Specifications

Antigen Cytokeratin 6
Clone LL020
Host Species Mouse
Species Reactivity Human
Applications Enzyme Linked Immunosorbent Assay
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122868

 
life technologies
LS14978880
25μg Each for $131.95

501122869

 
life technologies
LS14978882
100μg Each for $323.35
Description & Specifications

Specifications

Antigen Cytokeratin 6
Clone LL020
Host Species Mouse
Species Reactivity Human
Applications Enzyme Linked Immunosorbent Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody LL020 recognizes human Cytokeratin 6. Cytokeratins are intermediate filament proteins comprising one component of the cytoskeleton. Cytokeratin 6 is a type II (basic) keratin that associates with Cytokeratin 16 and Cytokeratin 17. Cytokeratin 6 is expressed in the epidermis, outer root sheath of hair follicles, oral mucosa, suprabasal cells of stratified epithelia, and glandular epithelia. Gene mutations result in inherited skin and hair disorders. Some reports suggest that LL020 may have some crossreactivity to Cytokeratin 5.