Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 6 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ Cytokeratin 6 Ab-1, Mouse Monoclonal Antibody.

$238.04 - $692.39

Specifications

Antigen Cytokeratin 6 Ab-1
Clone LHK6B
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS766P

 
thermo scientific™ lab vision™
MS766P
200μg/mL 1mL Each for $604.45

MS766P0

 
thermo scientific™ lab vision™
MS766P0
200μg/mL 100μL Each for $238.04

MS766P1

 
thermo scientific™ lab vision™
MS766P1
200μg/mL 500μL Each for $413.20

MS766P1ABX

 
thermo scientific™ lab vision™
MS766P1ABX
1mg/mL 100μL Each for $532.71

MS766PABX

 
thermo scientific™ lab vision™
MS766PABX
1mg/mL 200μL Each for $692.39
Description & Specifications

Specifications

Antigen Cytokeratin 6 Ab-1
Clone LHK6B
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen A synthetic peptide (GSSTIKYTTTS) from the C-terminus of human keratin 6
Isotype IgG2a

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ Cytokeratin 6 Ab-1, Mouse Monoclonal Antibody.

Keratins 6 and 16 are expressed in keratinocytes that are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.

Host Species: Mouse

Clone: LHK6B

Isotype: IgG2a

Species Reactivity: Human and Mouse. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide (GSSTIKYTTTS) from the C-terminus of human Cytokeratin 6

Molecular Weight: 56 kDa

Recommended for:

  • Immunohistochemistry (not qualified)

Note:

USA: RUO; Int'l: RUO