missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 6 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ Cytokeratin 6 Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS766P1ABX

 View more versions of this product

Catalog No. MS766P1ABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ Cytokeratin 6 Ab-1, Mouse Monoclonal Antibody.

Keratins 6 and 16 are expressed in keratinocytes that are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.

Host Species: Mouse

Clone: LHK6B

Isotype: IgG2a

Species Reactivity: Human and Mouse. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide (GSSTIKYTTTS) from the C-terminus of human Cytokeratin 6

Molecular Weight: 56 kDa

Recommended for:

  • Immunohistochemistry (not qualified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytokeratin 6 Ab-1
LHK6B
Unlabeled
Murine
IgG2a
RUO
Primary
Human,Murine
Monoclonal
1mg/mL
Concentrated
A synthetic peptide (GSSTIKYTTTS) from the C-terminus of human keratin 6
100μL
Cytoskeletal Protein
Monoclonal