missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody.

$376.59 - $1,704.08

Specifications

Antigen Cytokeratin 5/6 Ab-2
Clone D5/16 B4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS1814R7 Thermo Scientific™ Lab Vision™
MS1814R7
7mL Each for $992.63
MS1814S Thermo Scientific™ Lab Vision™
MS1814S
1mL Each for $1,704.08
MS1814S0 Thermo Scientific™ Lab Vision™
MS1814S0
100μL Each for $376.59
MS1814S1 Thermo Scientific™ Lab Vision™
MS1814S1
500μL Each for $1,559.35
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Cytokeratin 5/6 Ab-2, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification.

Host Species: Mouse

Clone: D5/16 B4

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Isolated cytokeratin 5

Molecular Weight: 58 kDa (Cytokeratin 5), 56 kDa (Cytokeratin 6)

Positive Control: Mesothelioma, Squamous Cell Ca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Description & Specifications

Specifications

Cytokeratin 5/6 Ab-2
Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
D5/16 B4
Monoclonal
Murine
Cytoskeletal Protein
Isolated cytokeratin 5