Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 19, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

$231.86 - $626.73

Specifications

Antigen Cytokeratin 19
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB9021P

 
thermo scientific™ lab vision™
RB9021P
Purified Ab with BSA and Azide 1mL Each for $626.73

RB9021P0

 
thermo scientific™ lab vision™
RB9021P0
Purified Ab with BSA and Azide 100µL Each for $231.86

RB9021P1

 
thermo scientific
RB9021P1
Purified Ab with BSA and Azide 500µL Each for $423.92

RB9021R7

 
thermo scientific™ lab vision™
RB9021R7
Ready-to-use for IHC (UVLP) 7mL Each for $332.67
Description & Specifications

Specifications

Antigen Cytokeratin 19
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen A synthetic peptide derived from C-terminal of human Keratin 19
Isotype IgG

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments with Thermo Scientific™ Cytokeratin 19, Rabbit Polyclonal Antibody.

Keratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminal of human Cytokeratin 19

Molecular Weight: 40 kDa

Positive Control: LS174T

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO