Please login to your online account to display your discounted pricing

Cytokeratin 19 Mouse anti-Human, eFluor 570, Clone: BA17, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $397.30

Specifications

Antigen Cytokeratin 19
Clone BA17
Host Species Mouse
Gene Alias CK19, Keratin 19
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123663

 
life technologies
LS41989880
25μg Each for $160.95

501123664

 
life technologies
LS41989882
100μg Each for $397.30
Description & Specifications

Specifications

Antigen Cytokeratin 19
Clone BA17
Host Species Mouse
Gene Alias CK19, Keratin 19
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This BA17 monoclonal antibody reacts with human cytokeratin 19 (CK19), a 44kDa type I (acidic) intermediate filament protein that lacks the non-alpha-helical tail domain present in other keratins. Cytokeratins form the intracellular cytoskeletal network that maintains the integrity and stability of cells and tissues. Cytokeratin 19 is expressed in a wide variety of simple and stratified epithelial tissue. Moreover, cytokeratin 19 expression can be induced by vitamin A, SV40 transformation, and cancer. A soluble form of cytokeratin 19 generated by caspase 3 cleavage has also been found to be secreted by cancer cells, thus possibly indicating tumor metastasis. Cytokeratin 19 often exists as a heterodimer with cytokeratin 7, a type II keratin.