missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Thermo Scientific™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.

$238.63 - $739.04

Specifications

Antigen Cytokeratin 19 Ab-5
Clone RCK108
Applications Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1902P Thermo Scientific™ Lab Vision™
MS1902P
Purified Ab with BSA and Azide 1mL Each for $651.43
MS1902P0 Thermo Scientific™ Lab Vision™
MS1902P0
Purified Ab with BSA and Azide 100μL Each for $238.63
MS1902P1 Thermo Scientific™ Lab Vision™
MS1902P1
Purified Ab with BSA and Azide 500μL Each for $437.35
MS1902P1ABX Thermo Scientific™ Lab Vision™
MS1902P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $563.86
MS1902PABX Thermo Scientific™ Lab Vision™
MS1902PABX
Ab without BSA/Azide (Frozen) 200μL Each for $739.04
MS1902R7 Thermo Scientific™ Lab Vision™
MS1902R7
Ready-to-use for IHC (UVLP) 7mL Each for $310.96
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Thermo Scientific™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: RCK108

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Cytoskeleton preparation of T24 cells

Molecular Weight: 40 kDa

Positive Control: LS174T and HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytokeratin 19 Ab-5
Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Unlabeled
RUO
Primary
IgG1
RCK108
Monoclonal
Murine
Cytoskeletal Protein
Cytoskeleton preparation of T24 cells