missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Thermo Scientific™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1902PABX

 View more versions of this product

Catalog No. MS1902PABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Thermo Scientific™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: RCK108

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Cytoskeleton preparation of T24 cells

Molecular Weight: 40 kDa

Positive Control: LS174T and HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytokeratin 19 Ab-5
Monoclonal
1mg/mL
Human
Ab without BSA/Azide (Frozen)
Cytoskeleton preparation of T24 cells
200μL
Cytoskeletal Protein
Monoclonal
Immunohistochemistry (Paraffin),Western Blotting
RCK108
Unlabeled
Concentrated
Murine
IgG1
RUO
Primary
Human