Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 19 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

$236.24 - $657.24

Specifications

Antigen Cytokeratin 19 Ab-1
Clone A53-B/A2.26; same as Ks19.1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Format Quantity Price Quantity    

MS198P

 
thermo scientific™ lab vision™
MS198P
200μg/mL Concentrated 1mL Each for $605.22

MS198P0

 
thermo scientific™ lab vision™
MS198P0
200μg/mL Concentrated 100µL Each for $236.24

MS198P1

 
thermo scientific™ lab vision™
MS198P1
200μg/mL Concentrated 500µL Each for $413.16

MS198P1ABX

 
thermo scientific™ lab vision™
MS198P1ABX
1mg/mL Concentrated 100µL Each for $510.94

MS198PABX

 
thermo scientific™ lab vision™
MS198PABX
1mg/mL Concentrated 200µL Each for $657.24

MS198R7

 
thermo scientific™ lab vision™
MS198R7
---- Prediluted 7mL Each for $333.55
Description & Specifications

Specifications

Antigen Cytokeratin 19 Ab-1
Clone A53-B/A2.26; same as Ks19.1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Host Species Mouse
Type Primary
Immunogen Human breast cancer MCF-7 cells
Isotype IgG2a lambda

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments with Thermo Scientific™ Cytokeratin 19 Ab-1, Mouse Monoclonal Antibody.

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues, excluding the liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: A53-B/A2.26; also known as Ks19.1

Isotype: IgG2a/λ

Species Reactivity: Human. Reacts weakly with Rat, Mouse and Guinea Pig. Others not known.

Epitope: aa 312-335

Immunogen: Human breast cancer MCF-7 cells

Molecular Weight: 40 kDa

Positive Control: HT29 and LS174T cells, skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence