missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunofluorescence applications with Thermo Scientific™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody.

$249.90 - $809.33

Specifications

Antigen Cytokeratin 15 Ab-1
Clone LHK15
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1068P Thermo Scientific™ Lab Vision™
MS1068P
200μg/mL 1mL Each for $723.76
MS1068P0 Thermo Scientific™ Lab Vision™
MS1068P0
200μg/mL 100μL Each for $249.90
MS1068P1 Thermo Scientific™ Lab Vision™
MS1068P1
200μg/mL 500μL Each for $564.39
MS1068P1ABX Thermo Scientific™ Lab Vision™
MS1068P1ABX
1mg/mL 100μL Each for $642.51
MS1068PABX Thermo Scientific™ Lab Vision™
MS1068PABX
1mg/mL 200μL Each for $809.33
Specifications

Description

Ensure accurate, reproducible results in immunofluorescence applications with Thermo Scientific™ Cytokeratin 15 Ab-1, Mouse Monoclonal Antibody.

Keratin 15 is a type I keratin without a defined type II partner. Keratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail. Expression of keratin 15 is downregulated in some hyperproliferating situations, such as psoriasis and hypertrophic scars. Because keratinocytes in psoriasis and hypertrophic scars are activated, it is suggested that keratin 15 expression is not compatible with keratinocyte activation and the keratin 15 gene is downregulated to maintain the activated phenotype.

Host Species: Mouse

Clone: LHK15

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytokeratin 15 Ab-1
Immunofluorescence
Unlabeled
Murine
Cytoskeletal Protein
A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15
LHK15
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa