Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

$225.69 - $963.20

Specifications

Antigen Cytokeratin 14 Ab-1
Clone LL002
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS115B

 
thermo scientific™ lab vision™
MS115B
Biotinylated Ab with BSA and Azide 1mL Each for $963.20

MS115B0

 
thermo scientific™ lab vision™
MS115B0
Biotinylated Ab with BSA and Azide 100μL Each for $314.41

MS115B1

 
thermo scientific™ lab vision™
MS115B1
Biotinylated Ab with BSA and Azide 500μL Each for $705.16

MS115P

 
thermo scientific™ lab vision™
MS115P
Purified Ab with BSA and Azide 1mL Each for $642.59

MS115P0

 
thermo scientific™ lab vision™
MS115P0
Purified Ab with BSA and Azide 100μL Each for $225.69

MS115P1

 
thermo scientific™ lab vision™
MS115P1
Purified Ab with BSA and Azide 500μL Each for $506.51

MS115P1ABX

 
thermo scientific™ lab vision™
MS115P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $590.96

MS115PABX

 
thermo scientific™ lab vision™
MS115PABX
Ab without BSA/Azide (Frozen) 200μL Each for $732.45

MS115R7

 
thermo scientific™ lab vision™
MS115R7
Ready-to-use for IHC (UVLP) 7mL Each for $330.65
Description & Specifications

Specifications

Antigen Cytokeratin 14 Ab-1
Clone LL002
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Host Species Murine
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14
Isotype IgG3

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.

Host Species: Mouse

Clone: LL002

Isotype: IgG3

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide of 15 amino acid residues from the C-terminus of human Cytokeratin 14

Molecular Weight: 50 kDa

Positive Control: A431 cells; Skin or Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO