Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS115P1ABX

 View more versions of this product

Catalog No. MS115P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 14 Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Clone LL002
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen A synthetic peptide of 15 amino acid residues from the C-terminus of human keratin 14
Isotype IgG3
Quantity 100μL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human
Target Species Rat

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Cytokeratin 14 Ab-1, Mouse Monoclonal Antibody.

Keratin 14 belongs to the type A (acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5. Keratin 14 has been studied as a prognostic marker in breast cancer.

Host Species: Mouse

Clone: LL002

Isotype: IgG3

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide of 15 amino acid residues from the C-terminus of human Cytokeratin 14

Molecular Weight: 50 kDa

Positive Control: A431 cells; Skin or Squamous cell CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO