Please login to your online account to display your discounted pricing

Alfa Aesar™ Cytochrome C, Equine Heart, Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67534LPL

 View more versions of this product

Catalog No. AAJ67534LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Cytochrome C
Quantity 1 vial
Storage Requirements -30°C to -10°C
CAS 9007-43-6
Format Lyophilized powder
Odor Odorless
Solubility Soluble in water at 100g/L
Description Dark red
For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.