Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytochrome c Ab-3, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in ELISA, flow cytometry and immunoprecipitation experiments with Thermo Scientific™ Cytochrome c Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1273P1ABX

 View more versions of this product

Catalog No. MS1273P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Host Species Mouse
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Immunogen Equine cytochrome c
Quantity 100µL
Antigen Cytochrome c Ab-3
Isotype IgG2a kappa
Clone CTC03; same as 2B5
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline Cancer and Tumor Biology

Obtain accurate, reproducible results in ELISA, flow cytometry and immunoprecipitation experiments with Thermo Scientific™ Cytochrome c Ab-3, Mouse Monoclonal Antibody.

Cytochrome c is an electron transporting protein that resides within the inter-membrane space of the mitochondria, where it plays a critical role in the process of oxidative phosphorylation and production of cellular ATP. An increasing amount of interest has been directed toward the role which cytocrome c has been demonstrated to play in apoptotic processes. Following exposure to apoptotic stimuli, cytochrome c is rapidly released from the mitochondria into the cytosol, an event which may be required for the completion of apoptosis in some systems. Cytosolic cytochrome c functions in the activation of caspase-3, and ICE family molecule that is a key effector of apoptosis.

Host Species: Mouse

Clone: CTC03; same as 2B5

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Horse and Dog. Others not known.

Epitope: Not determined

Immunogen: Equine cytochrome c

Molecular Weight: 15 kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting (Not suitable)
  • ELISA
  • Flow Cytometry
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO