missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytochrome c Ab-2, Mouse Monoclonal Antibody

Specifically detect Cytochrome c in human, mouse, rat, horse, dog, pigeon, frog and Drosophila samples with Thermo Scientific™ Cytochrome c Ab-2, Mouse Monoclonal Antibody. 

$248.96 - $1,242.49

Specifications

Antigen Cytochrome c Ab-2
Clone 7H8.2C12
Applications Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1192P Thermo Scientific™ Lab Vision™
MS1192P
Purified Ab with BSA and Azide 1mL Each for $1,156.12
MS1192P0 Thermo Scientific™ Lab Vision™
MS1192P0
Purified Ab with BSA and Azide 100μL Each for $248.96
MS1192P1 Thermo Scientific™ Lab Vision™
MS1192P1
Purified Ab with BSA and Azide 500μL Each for $765.29
MS1192P1ABX Thermo Scientific™ Lab Vision™
MS1192P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $849.71
MS1192PABX Thermo Scientific™ Lab Vision™
MS1192PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,242.49
MS1192R7 Thermo Scientific™ Lab Vision™
MS1192R7
Ready-to-use for IHC (UVLP) 7mL Each for $849.71
Specifications

Description

Specifically detect Cytochrome c in human, mouse, rat, horse, dog, pigeon, frog and Drosophila samples with Thermo Scientific™ Cytochrome c Ab-2, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

Cytochrome c is an electron transporting protein that resides within the inter-membrane space of the mitochondria, where it plays a critical role in the process of oxidative phosphorylation and production of cellular ATP. An increasing amount of interest has been directed toward the role which cytocrome c has been demonstrated to play in apoptotic processes. Following exposure to apoptotic stimuli, cytochrome c is rapidly released from the mitochondria into the cytosol, an event which may be required for the completion of apoptosis in some systems. Cytosolic cytochrome c functions in the activation of caspase-3, and ICE family molecule that is a key effector of apoptosis.

Host Species: Mouse

Clone: 7H8.2C12

Isotype: IgG2b/κ

Species Reactivity: Human, Mouse, Rat, Horse, Dog, Pigeon, Frog and Drosophila. Others not known.

Epitope: Not determined

Immunogen: Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon cytochrome c

Molecular Weight: 15 kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Cytochrome c Ab-2
Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Unlabeled
RUO
Primary
IgG2b kappa
7H8.2C12
Monoclonal
Murine
Apoptosis
Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon cytochrome c