Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cytochrome c Ab-2, Mouse Monoclonal Antibody


Specifically detect Cytochrome c in human, mouse, rat, horse, dog, pigeon, frog and Drosophila samples with Thermo Scientific™ Cytochrome c Ab-2, Mouse Monoclonal Antibody. 

Manufacturer: thermo scientific™ lab vision™  MS1192P1ABX

 View more versions of this product

Catalog No. MS1192P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Cytochrome c Ab-2
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Clone 7H8.2C12
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Drosophila
Cross Reactivity Equine
Cross Reactivity Frog
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Pigeon
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon cytochrome c
Isotype IgG2b kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Canine
Target Species Drosophila
Target Species Equine
Target Species Frog
Target Species Human
Target Species Murine
Target Species Pigeon
Target Species Rat

Specifically detect Cytochrome c in human, mouse, rat, horse, dog, pigeon, frog and Drosophila samples with Thermo Scientific™ Cytochrome c Ab-2, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

Cytochrome c is an electron transporting protein that resides within the inter-membrane space of the mitochondria, where it plays a critical role in the process of oxidative phosphorylation and production of cellular ATP. An increasing amount of interest has been directed toward the role which cytocrome c has been demonstrated to play in apoptotic processes. Following exposure to apoptotic stimuli, cytochrome c is rapidly released from the mitochondria into the cytosol, an event which may be required for the completion of apoptosis in some systems. Cytosolic cytochrome c functions in the activation of caspase-3, and ICE family molecule that is a key effector of apoptosis.

Host Species: Mouse

Clone: 7H8.2C12

Isotype: IgG2b/κ

Species Reactivity: Human, Mouse, Rat, Horse, Dog, Pigeon, Frog and Drosophila. Others not known.

Epitope: Not determined

Immunogen: Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon cytochrome c

Molecular Weight: 15 kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO